Tisk…

Azetidin-3-ylmethanol hydrochloride 97%

Dodavatel: Apollo Scientific

Synonyms: 3-Azetidinemethanol hydrochloride

OR309042-1G OR309042-5G
APOSOR309042-1GEA 2030 CZK
APOSOR309042-1G APOSOR309042-5G
Azetidin-3-ylmethanol hydrochloride 97%
Azetidin-3-ylmethanol hydrochloride
Vzorec: C₄H₁₀ClNO
MW: 123,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09743446
CAS číslo: 928038-44-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR