Tisk…

1-Benzyl 3-methyl azetidine-1,3-dicarboxylate 97%

Dodavatel: Apollo Scientific
Varování

OR309031-25G OR309031-5G
APOSOR309031-25GEA 5860 CZK
APOSOR309031-25G APOSOR309031-5G
1-Benzyl 3-methyl azetidine-1,3-dicarboxylate 97%
1-Benzyl 3-methyl azetidine-1,3-dicarboxylate
Vzorec: C₁₃H₁₅NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD11110708
CAS číslo: 757239-60-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR