1-Methyl-1H-indazol-5-ol 98%
Dodavatel: Apollo Scientific
OR30843-1G
OR30843-250MG
OR30843-5G
APOSOR30843-1GEA
7180
CZK
APOSOR30843-1G
APOSOR30843-250MG
APOSOR30843-5G
1-Methyl-1H-indazol-5-ol 98%
1-Methyl-1H-indazol-5-ol
Vzorec:
C₈H₈N₂O Storage Temperature: Ambient |
MDL Number:
MFCD11869778 CAS číslo: 756839-14-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...