Tisk…

5-Bromo-4-methyl-1H-indazole

Dodavatel: Apollo Scientific
Nebezpečí

OR30838-1G OR30838-5G OR30838-10G
APOSOR30838-1GEA 992 CZK
APOSOR30838-1G APOSOR30838-5G APOSOR30838-10G
5-Bromo-4-methyl-1H-indazole
5-Bromo-4-methyl-1H-indazole
Vzorec: C₈H₇BrN₂
Storage Temperature: Ambient
MDL Number: MFCD11007842
CAS číslo: 1082041-34-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR