(S)-3-Methylmorpholine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR308174-1G
OR308174-5G
OR308174-25G
APOSOR308174-1GEA
2030
CZK
APOSOR308174-1G
APOSOR308174-5G
APOSOR308174-25G
(S)-3-Methylmorpholine
(S)-3-Methylmorpholine
Vzorec:
C₅H₁₁NO MW: 101,1469 g/mol Bod varu: 133…136 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03840369 CAS číslo: 350595-57-2 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...