Tisk…

(S)-Morpholine-3-carboxylic acid

Dodavatel: Apollo Scientific

OR308162-1G OR308162-5G
APOSOR308162-1GEA 3580 CZK
APOSOR308162-1G APOSOR308162-5G
(S)-Morpholine-3-carboxylic acid
(S)-Morpholine-3-carboxylic acid
Vzorec: C₅H₉NO₃
Storage Temperature: Ambient
CAS číslo: 106825-79-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR