Tisk…

tert-Butyl methyl(piperidin-4-ylmethyl)carbamate

Dodavatel: Apollo Scientific

OR308129-1G OR308129-5G
APOSOR308129-1GEA 5290 CZK
APOSOR308129-1G APOSOR308129-5G
tert-Butyl methyl(piperidin-4-ylmethyl)carbamate
tert-Butyl methyl(piperidin-4-ylmethyl)carbamate
Vzorec: C₁₂H₂₄N₂O₂
MW: 228,34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06800387
CAS číslo: 138022-04-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR