tert-Butyl n-(3-methylpiperidin-3-yl)carbamate 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
tert-Butyl (3-methylpiperidin-3-yl)carbamate, 3-Amino-3-methylpiperidine, 3-BOC protected
OR308123-1G
OR308123-5G
APOSOR308123-1GEA
1660
CZK
APOSOR308123-1G
APOSOR308123-5G
tert-Butyl n-(3-methylpiperidin-3-yl)carbamate 97%
tert-Butyl n-(3-methylpiperidin-3-yl)carbamate
Vzorec:
C₁₁H₂₂N₂O₂ MW: 214.305 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD13194151 CAS číslo: 169750-96-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...