3-(Hydroxymethyl)cyclobutanone 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR308103-1G
OR308103-5G
APOSOR308103-1GEA
5230
CZK
APOSOR308103-1G
APOSOR308103-5G
3-(Hydroxymethyl)cyclobutanone 96%
3-(Hydroxymethyl)cyclobutanone
Vzorec:
C₅H₈O₂ MW: 100,12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11100969 CAS číslo: 183616-18-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...