Tisk…

3-Hydroxycyclobutanecarboxylic acid

Dodavatel: Apollo Scientific

OR308101-1G OR308101-5G
APOSOR308101-1GEA 1070 CZK
APOSOR308101-1G APOSOR308101-5G
3-Hydroxycyclobutanecarboxylic acid
3-Hydroxycyclobutanecarboxylic acid
Vzorec: C₅H₈O₃
Storage Temperature: Ambient
MDL Number: MFCD08705859
CAS číslo: 194788-10-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR