Tisk…

6-Quinazolinecarboxylic acid

Dodavatel: Apollo Scientific

OR308084-100MG OR308084-250MG
APOSOR308084-100MGEA 6290 CZK
APOSOR308084-100MG APOSOR308084-250MG
6-Quinazolinecarboxylic acid
6-Quinazolinecarboxylic acid
Vzorec: C₉H₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11520386
CAS číslo: 676326-53-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR