Tisk…

7H-Pyrrolo[2,3-d]pyrimidine-4-carboxylic acid

Dodavatel: Apollo Scientific

OR308079-1G OR308079-250MG
APOSOR308079-1GEA 4200 CZK
APOSOR308079-1G APOSOR308079-250MG
7H-Pyrrolo[2,3-d]pyrimidine-4-carboxylic acid
7H-Pyrrolo[2,3-d]pyrimidine-4-carboxylic acid
Vzorec: C₇H₅N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD13193623
CAS číslo: 1005206-17-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR