Tisk…

(S)-1-Boc-(2-hydroxymethyl)piperazine

Dodavatel: Apollo Scientific
Varování

OR308073-1G OR308073-5G
APOSOR308073-1GEA 3770 CZK
APOSOR308073-1G APOSOR308073-5G
(S)-1-Boc-(2-hydroxymethyl)piperazine
(S)-1-Boc-(2-hydroxymethyl)piperazine
Vzorec: C₁₀H₂₀N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD07772093
CAS číslo: 1030377-21-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR