Tisk…

(S)-1-tert-Butyl-2-methylpiperazine-1,2-dicarboxylate

Dodavatel: Apollo Scientific
Varování

Synonyms: (S)-1-tert-Butyl 2-methyl piperazine-1,2-dicarboxylate

OR308067-1G OR308067-5G
APOSOR308067-1GEA 2780 CZK
APOSOR308067-1G APOSOR308067-5G
(S)-1-tert-Butyl-2-methylpiperazine-1,2-dicarboxylate
(S)-1-tert-Butyl-2-methylpiperazine-1,2-dicarboxylate
Vzorec: C₁₁H₂₀N₂O₄
MW: 244,2875 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115328
CAS číslo: 796096-64-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR