Tisk…

2-Methylindazole-7-boronic acid pinacol ester

Dodavatel: Apollo Scientific

Synonyms: 2-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole

OR308059-1G OR308059-5G
APOSOR308059-1GEA 7250 CZK
APOSOR308059-1G APOSOR308059-5G
2-Methylindazole-7-boronic acid pinacol ester
2-Methylindazole-7-boronic acid pinacol ester
Vzorec: C₁₄H₁₉BN₂O₂
MW: 258.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11109457
CAS číslo: 845751-67-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR