Tisk…

2-(1-tert-Butoxycarbonylazetidin-3-yl)acetic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-[1-Boc-azetidin-3-yl]acetic acid, 2-[1-(tert-Butoxycarbonyl)azetidin-3-yl]acetic acid

OR308024-25G
APOSOR308024-25GEA 2570 CZK
APOSOR308024-25G
2-(1-tert-Butoxycarbonylazetidin-3-yl)acetic acid
2-(1-tert-Butoxycarbonylazetidin-3-yl)acetic acid
Vzorec: C₁₀H₁₇NO₄
MW: 215,25 g/mol
Bod varu: 341,6 °C/ 760mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD02179028
CAS číslo: 183062-96-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR