Tisk…

Oxetan-3-ylmethanol 96%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-(Hydroxymethyl)oxetane, 3-Oxetanemethanol, Oxetan-3-ylmethanol

OR308006-1G OR308006-5G
APOSOR308006-1GEA 1160 CZK
APOSOR308006-1G APOSOR308006-5G
Oxetan-3-ylmethanol 96%
Oxetan-3-ylmethanol
Vzorec: C₄H₈O₂
MW: 88,11 g/mol
Storage Temperature: Freezer
MDL Number: MFCD17015894
CAS číslo: 6246-06-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR