4-Chloro-7,8-dimethylquinoline
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR307968-1G
OR307968-5G
APOSOR307968-1GEA
5030
CZK
APOSOR307968-1G
APOSOR307968-5G
4-Chloro-7,8-dimethylquinoline
4-Chloro-7,8-dimethylquinoline
Vzorec:
C₁₁H₁₀ClN Bod tání: 61…62 °C Storage Temperature: Ambient |
CAS číslo:
181950-53-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...