4-Chloro-5,7-dimethylquinoline
Dodavatel: Apollo Scientific
OR307963-1G
OR307963-5G
APOSOR307963-1GEA
8570
CZK
APOSOR307963-1G
APOSOR307963-5G
4-Chloro-5,7-dimethylquinoline
4-Chloro-5,7-dimethylquinoline
Vzorec:
C₁₁H₁₀ClN Storage Temperature: Ambient |
CAS číslo:
352205-97-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...