Tisk…

4-Chloro-2,7,8-trimethylquinoline

Dodavatel: Apollo Scientific

OR307962-1G OR307962-5G
APOSOR307962-1GEA 8570 CZK
APOSOR307962-1G APOSOR307962-5G
4-Chloro-2,7,8-trimethylquinoline
4-Chloro-2,7,8-trimethylquinoline
Vzorec: C₁₂H₁₂ClN
Storage Temperature: Ambient
CAS číslo: 78509-29-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR