4-Amino-6-bromoquinoline
Dodavatel: Apollo Scientific
OR307942-1G
OR307942-5G
APOSOR307942-1GEA
3380
CZK
APOSOR307942-1G
APOSOR307942-5G
4-Amino-6-bromoquinoline
4-Amino-6-bromoquinoline
Vzorec:
C₉H₇BrN₂ MW: 223,0693 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03407953 CAS číslo: 65340-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...