meso-tert-Butyl hexahydropyrrolo[3,4-c]pyrrole-2(1H)-carboxylate 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
cis-3,7-Diazabicyclo[3.3.0]octane, N3-BOC protected, cis-2-Boc-Hexahydropyrrol[3,4-c]pyrrole
OR30794-1G
OR30794-250MG
APOSOR30794-1GEA
3060
CZK
APOSOR30794-1G
APOSOR30794-250MG
meso-tert-Butyl hexahydropyrrolo[3,4-c]pyrrole-2(1H)-carboxylate 97%
meso-tert-Butyl hexahydropyrrolo[3,4-c]pyrrole-2(1H)-carboxylate
Vzorec:
C₁₁H₂₀N₂O₂ MW: 212,29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04116224 CAS číslo: 250275-15-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...