Tisk…

4,7-Dichloro-2,8-dimethylquinoline

Dodavatel: Apollo Scientific

OR307933-5G
APOSOR307933-5GEA 4630 CZK
APOSOR307933-5G
4,7-Dichloro-2,8-dimethylquinoline
4,7-Dichloro-2,8-dimethylquinoline
Vzorec: C₁₁H₉Cl₂N
MW: 226,1000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272342
CAS číslo: 21728-15-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR