3-Bromo-2-hydroxybenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR30793-1G
OR30793-5G
OR30793-250MG
APOSOR30793-1GEA
705
CZK
APOSOR30793-1G
APOSOR30793-5G
APOSOR30793-250MG
3-Bromo-2-hydroxybenzoic acid
3-Bromo-2-hydroxybenzoic acid
Vzorec:
C7H5BrO3 Storage Temperature: Refrigerator |
MDL Number:
MFCD03265705 CAS číslo: 3883-95-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...