2-Chloro-5,7-dimethylquinoline-3-carbonitrile
Dodavatel: Apollo Scientific
OR307911-1G
OR307911-5G
APOSOR307911-1GEA
8570
CZK
APOSOR307911-1G
APOSOR307911-5G
2-Chloro-5,7-dimethylquinoline-3-carbonitrile
2-Chloro-5,7-dimethylquinoline-3-carbonitrile
Vzorec:
C₁₂H₉ClN₂ Storage Temperature: Ambient |
MDL Number:
MFCD08899072 CAS číslo: 917747-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...