2-Chloro-3,8-dimethylquinoline
Dodavatel: Apollo Scientific
OR307903-1G
OR307903-5G
APOSOR307903-1GEA
8340
CZK
APOSOR307903-1G
APOSOR307903-5G
2-Chloro-3,8-dimethylquinoline
2-Chloro-3,8-dimethylquinoline
Vzorec:
C₁₁H₁₀ClN Storage Temperature: Ambient |
CAS číslo:
108097-04-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...