Tisk…

2,7,8-Trimethyl-4-quinolinol

Dodavatel: Apollo Scientific

Synonyms: 4-Hydroxy-2,7,8-trimethylquinoline

OR307887-1G OR307887-5G
APOSOR307887-1GEA 4300 CZK
APOSOR307887-1G APOSOR307887-5G
2,7,8-Trimethyl-4-quinolinol
2,7,8-Trimethyl-4-quinolinol
Vzorec: C₁₂H₁₃NO
Storage Temperature: Ambient
CAS číslo: 449199-19-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR