Tisk…

2,5,7-Trimethylquinoline-3-carboxylic acid ethyl ester

Dodavatel: Apollo Scientific

OR307878-1G OR307878-5G
APOSOR307878-1GEA 8340 CZK
APOSOR307878-1G APOSOR307878-5G
2,5,7-Trimethylquinoline-3-carboxylic acid ethyl ester
2,5,7-Trimethylquinoline-3-carboxylic acid ethyl ester
Vzorec: C₁₅H₁₇NO₂
Storage Temperature: Ambient
CAS číslo: 948291-02-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR