2,5,7-Trimethylquinoline
Dodavatel: Apollo Scientific
OR307877-1G
OR307877-5G
APOSOR307877-1GEA
8340
CZK
APOSOR307877-1G
APOSOR307877-5G
2,5,7-Trimethylquinoline
2,5,7-Trimethylquinoline
Vzorec:
C₁₂H₁₃N Storage Temperature: Ambient |
CAS číslo:
102871-67-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...