4-Amino-5,7-dichloro-2-methylquinoline
Dodavatel: Apollo Scientific
OR307862-1G
OR307862-5G
APOSOR307862-1GEA
7870
CZK
APOSOR307862-1G
APOSOR307862-5G
4-Amino-5,7-dichloro-2-methylquinoline
4-Amino-5,7-dichloro-2-methylquinoline
Vzorec:
C₁₀H₈Cl₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD08669723 CAS číslo: 917562-03-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...