Tisk…

2,4-Dichloro-6-methylpyridine

Dodavatel: Apollo Scientific
Varování

OR30785-1G OR30785-5G
APOSOR30785-1GEA 744 CZK
APOSOR30785-1G APOSOR30785-5G
2,4-Dichloro-6-methylpyridine
2,4-Dichloro-6-methylpyridine
Vzorec: C₆H₅Cl₂N
MW: 162,01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08669834
CAS číslo: 42779-56-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR