Tisk…

5,7-Dimethylquinolin-4(1H)-one

Dodavatel: Apollo Scientific

Synonyms: 5,7-Dimethyl-4-hydroxyquinoline

OR307843-1G OR307843-5G
APOSOR307843-1GEA 8340 CZK
APOSOR307843-1G APOSOR307843-5G
5,7-Dimethylquinolin-4(1H)-one
5,7-Dimethylquinolin-4(1H)-one
Vzorec: C₁₁H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD00272455
CAS číslo: 203626-56-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR