8-Chloroquinoline-2,3-dicarboxylic acid diethyl ester
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR307841-1G
OR307841-5G
APOSOR307841-1GEA
6220
CZK
APOSOR307841-1G
APOSOR307841-5G
8-Chloroquinoline-2,3-dicarboxylic acid diethyl ester
8-Chloroquinoline-2,3-dicarboxylic acid diethyl ester
Vzorec:
C₁₅H₁₄ClNO₄ Bod tání: 106…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD08437572 CAS číslo: 892874-60-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...