Tisk…
7-Chloroquinoline-2,3-dicarboxylic acid diethyl ester
Dodavatel: Apollo Scientific
OR307838-1G OR307838-5G
APOSOR307838-1GEA 8340 CZK
APOSOR307838-1G APOSOR307838-5G
7-Chloroquinoline-2,3-dicarboxylic acid diethyl ester
7-Chloroquinoline-2,3-dicarboxylic acid diethyl ester
Vzorec: C₁₅H₁₄ClNO₄
Storage Temperature: Ambient
MDL Number: MFCD08437570
CAS číslo: 892874-55-4