Tisk…

5-(4-Morpholinyl)-2-nitroaniline

Dodavatel: Apollo Scientific

OR30783-1G OR30783-5G
APOSOR30783-1GEA 2690 CZK
APOSOR30783-1G APOSOR30783-5G
5-(4-Morpholinyl)-2-nitroaniline
5-(4-Morpholinyl)-2-nitroaniline
Vzorec: C₁₀H₁₃N₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00511844
CAS číslo: 54998-00-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR