7-Bromoquinoline-2,3-dicarboxylic acid diethyl ester
Dodavatel: Apollo Scientific
OR307823-1G
OR307823-5G
APOSOR307823-1GEA
8570
CZK
APOSOR307823-1G
APOSOR307823-5G
7-Bromoquinoline-2,3-dicarboxylic acid diethyl ester
7-Bromoquinoline-2,3-dicarboxylic acid diethyl ester
Vzorec:
C₁₅H₁₄BrNO₄ Storage Temperature: Ambient |
MDL Number:
MFCD08437558 CAS číslo: 892874-41-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...