Tisk…

8-Bromo-4-chloro-2-methylquinoline

Dodavatel: Apollo Scientific
Nebezpečí

OR307816-1G OR307816-5G
APOSOR307816-1GEA 2820 CZK
APOSOR307816-1G APOSOR307816-5G
8-Bromo-4-chloro-2-methylquinoline
8-Bromo-4-chloro-2-methylquinoline
Vzorec: C₁₀H₇BrClN
MW: 256,5300 g/mol
Bod tání: 73…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00272432
CAS číslo: 1201-07-6
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR