Tisk…

2-Amino-4-oxazolecarboxylic acid

Dodavatel: Apollo Scientific

OR30773-1G OR30773-250MG OR30773-500MG OR30773-2.5G OR30773-5G
APOSOR30773-1GEA 7080 CZK
APOSOR30773-1G APOSOR30773-250MG APOSOR30773-500MG APOSOR30773-2.5G APOSOR30773-5G
2-Amino-4-oxazolecarboxylic acid
2-Amino-4-oxazolecarboxylic acid
Vzorec: C₄H₄N₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD09864918
CAS číslo: 944900-52-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR