Tisk…

(1S,2R)-2-Aminocyclopentanol hydrochloride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: (1S,2R)-2-Aminocyclopentanol Hydrochloride

OR307678-1G
APOSOR307678-1GEA 1740 CZK
APOSOR307678-1G
(1S,2R)-2-Aminocyclopentanol hydrochloride
(1S,2R)-2-Aminocyclopentanol hydrochloride
Vzorec: C₅H₁₂ClNO
MW: 137,61 g/mol
Bod tání: 228…229 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD08704797
CAS číslo: 225791-13-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR