Tisk…

(1S,2R)-Boc-2-amino-1-cyclopentanecarboxylic acid

Dodavatel: Apollo Scientific
Varování

OR307675-1G OR307675-250MG
APOSOR307675-1GEA 3910 CZK
APOSOR307675-1G APOSOR307675-250MG
(1S,2R)-Boc-2-amino-1-cyclopentanecarboxylic acid
(1S,2R)-Boc-2-amino-1-cyclopentanecarboxylic acid
Vzorec: C₁₁H₁₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD09750528
CAS číslo: 137170-89-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR