Tisk…

5-Phenylhexahydro-4,7-methano-2-benzofuran-1,3-dione

Dodavatel: Apollo Scientific

OR307553-1G OR307553-250MG
APOSOR307553-1GEA 16110 CZK
APOSOR307553-1G APOSOR307553-250MG
5-Phenylhexahydro-4,7-methano-2-benzofuran-1,3-dione
5-Phenylhexahydro-4,7-methano-2-benzofuran-1,3-dione
Vzorec: C₁₅H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD13873730
CAS číslo: 73252-09-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR