4-Bromo-7-chloroquinoline
Dodavatel: Apollo Scientific
OR307507-5G
APOSOR307507-5GEA
3440
CZK
APOSOR307507-5G
4-Bromo-7-chloroquinoline
4-Bromo-7-chloroquinoline
Vzorec:
C₉H₅BrClN MW: 242,5000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07700232 CAS číslo: 98519-65-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...