Tisk…

4-Chloro-1H-pyrrolo[3,2-c]pyridine 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Chloro-5-azaindole

OR30747-5G OR30747-10G
APOSOR30747-5GEA 4900 CZK
APOSOR30747-5G APOSOR30747-10G
4-Chloro-1H-pyrrolo[3,2-c]pyridine 95%
4-Chloro-1H-pyrrolo[3,2-c]pyridine
Vzorec: C₇H₅ClN₂
MW: 152,5810 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07781160
CAS číslo: 60290-21-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR