6-Bromo-3-methyl-1H-indazole
Dodavatel: Apollo Scientific
|
Varování
|
OR30741-1G
OR30741-250MG
OR30741-5G
APOSOR30741-1GEA
2110
CZK
APOSOR30741-1G
APOSOR30741-250MG
APOSOR30741-5G
6-Bromo-3-methyl-1H-indazole
6-Bromo-3-methyl-1H-indazole
Vzorec:
C₈H₇BrN₂ MW: 211,0600 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08272244 CAS číslo: 7746-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...