Tisk…

1H-Indazole-6-boronic acid pinacol ester 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

OR30736-1G OR30736-5G
APOSOR30736-1GEA 1240 CZK
APOSOR30736-1G APOSOR30736-5G
1H-Indazole-6-boronic acid pinacol ester 97%
1H-Indazole-6-boronic acid pinacol ester
Vzorec: C₁₃H₁₇BN₂O₂
MW: 244.0973 g/mol
Bod tání: 134…140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368067
CAS číslo: 937049-58-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR