Tisk…

1H-Indazole-5-boronic acid pinacol ester

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

OR30735-1G OR30735-500MG
APOSOR30735-1GEA 1990 CZK
APOSOR30735-1G APOSOR30735-500MG
1H-Indazole-5-boronic acid pinacol ester
1H-Indazole-5-boronic acid pinacol ester
Vzorec: C₁₃H₁₇BN₂O₂
MW: 244.0973 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367523
CAS číslo: 862723-42-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR