Tisk…

5-Bromo-3-methyl-1H-indazole 98%

Dodavatel: Apollo Scientific
Varování

OR30734-1G OR30734-5G OR30734-10G
APOSOR30734-1GEA 622 CZK
APOSOR30734-1G APOSOR30734-5G APOSOR30734-10G
5-Bromo-3-methyl-1H-indazole 98%
5-Bromo-3-methyl-1H-indazole
Vzorec: C₈H₇BrN₂
MW: 211,0600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05664003
CAS číslo: 552331-16-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR