Tisk…

(1H-Indazol-5-yl)methanol 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-(Hydroxymethyl)-1H-indazole

OR30731-1G OR30731-5G
APOSOR30731-1GEA 2030 CZK
APOSOR30731-1G APOSOR30731-5G
(1H-Indazol-5-yl)methanol 95%
(1H-Indazol-5-yl)methanol
Vzorec: C₈H₈N₂O
MW: 148,1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038636
CAS číslo: 478828-52-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR