Tisk…

1H-Indazol-7-ol

Dodavatel: Apollo Scientific
Varování

Synonyms: 7-Hydroxy-1H-indazole

OR30730-1G OR30730-5G OR30730-10G
APOSOR30730-1GEA 787 CZK
APOSOR30730-1G APOSOR30730-5G APOSOR30730-10G
1H-Indazol-7-ol
1H-Indazol-7-ol
Vzorec: C₇H₆N₂O
MW: 134,1400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00464298
CAS číslo: 81382-46-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR