Tisk…

1-(Tetrahydropyranyl)-1H-indazole-5-carboxylic acid

Dodavatel: Apollo Scientific
Varování

OR30728-100MG OR30728-250MG
APOSOR30728-100MGEA 1210 CZK
APOSOR30728-100MG APOSOR30728-250MG
1-(Tetrahydropyranyl)-1H-indazole-5-carboxylic acid
1-(Tetrahydropyranyl)-1H-indazole-5-carboxylic acid
Vzorec: C₁₃H₁₄N₂O₃
MW: 246,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09870060
CAS číslo: 1000576-28-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR